2-Fluoro-5-nitrobenzotrifluoride ≥98.0%

Supplier: TCI America
Warning

F0373-25G F0373-5G
TCF0373-025GEA 145.52 CAD
TCF0373-025G TCF0373-005G
2-Fluoro-5-nitrobenzotrifluoride ≥98.0%
2-Fluoro-5-nitrobenzotrifluoride

CAS Number: 400-74-8
MDL Number: MFCD00024576
Molecular Formula: C7H3F4NO2
Molecular Weight: 209.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 91
Specific Gravity (20/20): 1.53


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Fluoro-4-nitro-2-(trifluoromethyl)benzene, 5-Nitro-alpha,alpha,alpha,2-tetrafluorotoluene

Formula: C₇H₃F₄NO₂
Boiling Pt: 110 °C
Melting Pt: 22…24 °C
Density: 1.53 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00024576
CAS Number: 400-74-8
EINECS: 206-925-8
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3b33e456c0056804ddfda28c40ac7ea843f0210&vid=e9314bb06bc49777d6c2e5314df4a6d58261d6ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR