3-Fluorobenzyl alcohol ≥97.0%
Supplier: TCI America
F0370-25G
F0370-5G
TCF0370-025GEA
237
CAD
TCF0370-025G
TCF0370-5G
3-Fluorobenzyl alcohol ≥97.0%
3-Fluorobenzyl alcohol
CAS Number: 456-47-3
MDL Number: MFCD00004631
Molecular Formula: C7H7FO
Molecular Weight: 126.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 201
Flash Point (°C): 90
Specific Gravity (20/20): 1.17
Formula:
C₇H₇FO Boiling Pt: 201 °C Density: 1.17 (20 °C) Flash Pt: 90 °C |
MDL Number:
MFCD00004631 CAS Number: 456-47-3 EINECS: 207-265-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...