5-Fluoroindole ≥98.0%

Supplier: TCI America
Warning

Synonyms: 5-Fluoro-1H-indole

F0369-1G F0369-5G
TCF0369-001GEA 55.3 CAD
TCF0369-001G TCF0369-005G
5-Fluoroindole ≥98.0%
5-Fluoroindole

CAS Number: 399-52-0
MDL Number: MFCD00005671
Molecular Formula: C8H6FN
Molecular Weight: 135.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 47
Flash Point (°C): 110

Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 120 °C
Melting Pt: 47 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005671
CAS Number: 399-52-0
EINECS: 206-917-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f55af34532e09d88fd6d7fd1b300de2e05f24b9e&vid=f55af34532e09d88fd6d7fd1b300de2e05f24b9e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR