3'-Fluoro-4'-methoxyacetophenone ≥98.0%
Supplier: TCI America
F0366-5G
F0366-25G
TCF0366-005GEA
80.58
CAD
TCF0366-005G
TCF0366-25G
3'-Fluoro-4'-methoxyacetophenone ≥98.0%
3'-Fluoro-4'-methoxyacetophenone
CAS Number: 455-91-4
MDL Number: MFCD00026219
Molecular Formula: C9H9FO2
Molecular Weight: 168.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 148
Melting point (°C): 92
Formula:
C₉H₉FO₂ Boiling Pt: 148 °C Melting Pt: 92 °C |
MDL Number:
MFCD00026219 CAS Number: 455-91-4 EINECS: 207-253-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...