6-Fluoroindole ≥98.0%

Supplier: TCI America
Warning

Synonyms: 6-Fluoro-1H-indole

F0352-1G F0352-5G
TCF0352-001GEA 62.88 CAD
TCF0352-001G TCF0352-005G
6-Fluoroindole ≥98.0%
6-Fluoroindole

CAS Number: 399-51-9
MDL Number: MFCD00056933
Molecular Formula: C8H6FN
Molecular Weight: 135.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 76

Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00056933
CAS Number: 399-51-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0b850497a8b0b05595de2e476058d2e696d0714&vid=c0b850497a8b0b05595de2e476058d2e696d0714&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR