3-Fluorophthalic anhydride ≥98.0%
Supplier: TCI America
F0349-5G
F0349-25G
TCF0349-005GEA
225.94
CAD
TCF0349-005G
TCF0349-25G
3-Fluorophthalic anhydride ≥98.0%
3-Fluorophthalic anhydride
CAS Number: 652-39-1
MDL Number: MFCD00039696
Molecular Formula: C8H3FO3
Molecular Weight: 166.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 283
Melting point (°C): 161
Formula:
C₈H₃FO₃ Boiling Pt: 283 °C Melting Pt: 161 °C |
MDL Number:
MFCD00039696 CAS Number: 652-39-1 EINECS: 211-491-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...