5'-Fluoro-2'-hydroxyacetophenone ≥98.0%

Supplier: TCI America

Synonyms: 1-(5-Fluoro-2-hydroxyphenyl)ethanone

F0318-5G F0318-25G
TCF0318-005GEA 56.88 CAD
TCF0318-005G TCF0318-025G
5'-Fluoro-2'-hydroxyacetophenone ≥98.0%
5'-Fluoro-2'-hydroxyacetophenone

CAS Number: 394-32-1
MDL Number: MFCD00011668
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 100
Melting point (°C): 57

Formula: C₈H₇FO₂
Boiling Pt: 100 °C
Melting Pt: 57 °C
MDL Number: MFCD00011668
CAS Number: 394-32-1
EINECS: 206-893-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6942d3f2fa13fda7eed9d34bc426cc854cef4e95&vid=6942d3f2fa13fda7eed9d34bc426cc854cef4e95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR