4-Fluoro-2-(trifluoromethyl)benzonitrile ≥98.0%

Supplier: TCI America

F0309-5G F0309-1G
TCF0309-5GEA 77.42 CAD
TCF0309-5G TCF0309-001G
4-Fluoro-2-(trifluoromethyl)benzonitrile ≥98.0%
4-Fluoro-2-(trifluoromethyl)benzonitrile

CAS Number: 194853-86-6
MDL Number: MFCD00061283
Molecular Formula: C8H3F4N
Molecular Weight: 189.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 46

Formula: C₈H₃F₄N
Boiling Pt: 52…55 °C (15 mmHg)
Melting Pt: 46 °C
Density: 1.35 g/cm³ (20 °C)
MDL Number: MFCD00061283
CAS Number: 194853-86-6
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6b0bec4c4961fa88145a5186945ee95918fc555&vid=e6b0bec4c4961fa88145a5186945ee95918fc555&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR