2-Fluoro-5-(trifluoromethyl)benzaldehyde ≥97.0%

Supplier: TCI America

F0289-5G F0289-1G F0289-25G
TCF0289-005GEA 241.74 CAD
TCF0289-005G TCF0289-001G TCF0289-25G
2-Fluoro-5-(trifluoromethyl)benzaldehyde ≥97.0%
2-Fluoro-5-(trifluoromethyl)benzaldehyde

CAS Number: 146137-78-2
MDL Number: MFCD00061311
Molecular Formula: C8H4F4O
Molecular Weight: 192.11
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.41

Formula: C₈H₄F₄O
Density: 1.41 (20 °C)
MDL Number: MFCD00061311
CAS Number: 146137-78-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dd52268a4a22942d8287927053713afd49a05c8&vid=2dd52268a4a22942d8287927053713afd49a05c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR