4-Fluorobenzamide ≥98.0% (by total nitrogen basis)
Supplier: TCI America
F0279-25G
F0279-5G
TCF0279-025GEA
186.44
CAD
TCF0279-025G
TCF0279-005G
4-Fluorobenzamide ≥98.0% (by total nitrogen basis)
4-Fluorobenzamide
CAS Number: 824-75-9
MDL Number: MFCD00007992
Molecular Formula: C7H6FNO
Molecular Weight: 139.13
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 155
Formula:
C₇H₆FNO Boiling Pt: 287 °C (1013 hPa) Melting Pt: 155 °C Density: 1.43 g/cm³ (20 °C) |
MDL Number:
MFCD00007992 CAS Number: 824-75-9 EINECS: 212-535-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...