2-Fluoro-5-nitrotoluene ≥97.0%

Supplier: TCI America

F0276-25G F0276-10G
TCF0276-25GEA 44.24 CAD
TCF0276-25G TCF0276-010G
2-Fluoro-5-nitrotoluene ≥97.0%
2-Fluoro-5-nitrotoluene

CAS Number: 455-88-9
MDL Number: MFCD00007284
Molecular Formula: C7H6FNO2
Molecular Weight: 155.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 99
Melting point (°C): 39
Flash Point (°C): 105
Storage Temperature: 0-10°C

Formula: C₇H₆FNO₂
Boiling Pt: 99 °C
Melting Pt: 39 °C
Flash Pt: 105 °C
MDL Number: MFCD00007284
CAS Number: 455-88-9
EINECS: 207-251-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a6e835855bb4ec2b305ec048ec456996ecc9f8a&vid=b33e2e9310d12bd710c1367c0eecefc6593b01ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR