3-Fluorobenzamide ≥97.0% (by total nitrogen basis)

Supplier: TCI America

F0266-5G F0266-25G
TCF0266-005GEA 109.02 CAD
TCF0266-005G TCF0266-025G
3-Fluorobenzamide ≥97.0% (by total nitrogen basis)
3-Fluorobenzamide

CAS Number: 455-37-8
MDL Number: MFCD00007983
Molecular Formula: C7H6FNO
Molecular Weight: 139.13
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 131

Formula: C₇H₆FNO
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 131 °C
MDL Number: MFCD00007983
CAS Number: 455-37-8
EINECS: 207-247-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ccdc0460bd3ad924f835e1942eba42d0c9d57b5b&vid=ccdc0460bd3ad924f835e1942eba42d0c9d57b5b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR