2-Fluorobiphenyl ≥97.0%

Supplier: TCI America

F0265-25G F0265-5G
TCF0265-25GEA 398.16 CAD
TCF0265-25G TCF0265-005G
2-Fluorobiphenyl ≥97.0%
2-Fluorobiphenyl

CAS Number: 321-60-8
MDL Number: MFCD00000317
Molecular Formula: C12H9F
Molecular Weight: 172.20
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 248
Melting point (°C): 74

Formula: C₁₂H₉F
Boiling Pt: 248 °C
Melting Pt: 74 °C
Density: 1.2452 g/cm³ (25 °C)
MDL Number: MFCD00000317
CAS Number: 321-60-8
EINECS: 206-290-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bb3ebe69c9b7296d32577b4204882f991cfbba7&vid=7bb3ebe69c9b7296d32577b4204882f991cfbba7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR