1-Fluoro-2-iodobenzene ≥99.0%

Supplier: TCI America

Synonyms: 2-Fluoroiodobenzene

F0253-10G F0253-25G
TCF0253-010GEA 31.6 CAD
TCF0253-010G TCF0253-025G
1-Fluoro-2-iodobenzene ≥99.0%
1-Fluoro-2-iodobenzene

CAS Number: 348-52-7
MDL Number: MFCD00001031
Molecular Formula: C6H4FI
Molecular Weight: 222.00
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 189
Flash Point (°C): 71
Specific Gravity (20/20): 1.93


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoroiodobenzene

Formula: C₆H₄FI
MW: 222 g/mol
Boiling Pt: 189 °C
Melting Pt: –41.5 °C
Density: 1.93 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00001031
CAS Number: 348-52-7
EINECS: 206-477-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cc6d81bd15580b3acfb22d4f88f59daec1313a4&vid=49a1e16851f5d1da46b240d41323c45ec14a4dd9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR