4-Fluorobenzyl alcohol ≥98.0%
Supplier: TCI America
F0251-25G
F0251-5G
TCF0251-025GEA
69.98
CAD
TCF0251-025G
TCF0251-5G
4-Fluorobenzyl alcohol ≥98.0%
4-Fluorobenzyl alcohol
CAS Number: 459-56-3
MDL Number: MFCD00004651
Molecular Formula: C7H7FO
Molecular Weight: 126.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 99
Flash Point (°C): 90
Freezing point (°C): 22
Formula:
C₇H₇FO Boiling Pt: 99 °C Melting Pt: 22…23 °C Density: 1.15 g/cm³ (20 °C) Flash Pt: 90 °C |
MDL Number:
MFCD00004651 CAS Number: 459-56-3 EINECS: 207-292-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...