4-Fluorocinnamic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0244-25G F0244-5G
TCF0244-025GEA 148.84 CAD
TCF0244-025G TCF0244-005G
4-Fluorocinnamic acid ≥98.0% (by GC, titration analysis)
4-Fluorocinnamic acid

CAS Number: 459-32-5
MDL Number: MFCD00004395
Molecular Formula: C9H7FO2
Molecular Weight: 166.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 207

Formula: C₉H₇FO₂
Melting Pt: 207 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00004395
CAS Number: 459-32-5
EINECS: 207-288-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5adf12edc826c7606c91fb88b8b58c858a60371&vid=a5adf12edc826c7606c91fb88b8b58c858a60371&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR