Methyl-2-fluorobenzoate ≥98.0%
Supplier: TCI America
F0241-25G
F0241-500G
TCF0241-025GEA
48.98
CAD
TCF0241-025G
TCF0241-500G
Methyl-2-fluorobenzoate ≥98.0%
Methyl 2-fluorobenzoate
CAS Number: 394-35-4
MDL Number: MFCD00017913
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 94
Specific Gravity (20/20): 1.21
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluorobenzoic Acid Methyl Ester
Formula:
C₈H₇FO₂ Boiling Pt: 102 °C Density: 1.21 (20 °C) Flash Pt: 94 °C |
MDL Number:
MFCD00017913 CAS Number: 394-35-4 EINECS: 206-894-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...