1-Fluoro-4-iodobenzene ≥98.0% stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with Stabilised

F0237-5G F0237-25G
TCF0237-005GEA 36.34 CAD
TCF0237-005G TCF0237-025G
1-Fluoro-4-iodobenzene ≥98.0% stabilized
1-Fluoro-4-iodobenzene

(stabilized with Copper chip)
CAS Number: 352-34-1
MDL Number: MFCD00001052
Molecular Formula: C6H4FI
Molecular Weight: 222.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 183
Melting point (°C): -20
Flash Point (°C): 68
Specific Gravity (20/20): 1.92


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 4-Fluoroiodobenzene

Formula: C₆H₄FI
Boiling Pt: 183 °C
Melting Pt: –20 °C
Density: 1.92 (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00001052
CAS Number: 352-34-1
EINECS: 206-522-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdfc36d0c8db0cf142c5a90f082c6d2575d29ea0&vid=b9f34a370c474dc6e7fcd5a01a05ed93c06f6506&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR