4-Fluoro-2-nitrotoluene ≥96.0%

Supplier: TCI America
Warning

F0218-25G F0218-5G
TCF0218-025GEA 84.37 CAD
TCF0218-025G TCF0218-005G
4-Fluoro-2-nitrotoluene ≥96.0%
4-Fluoro-2-nitrotoluene

CAS Number: 446-10-6
MDL Number: MFCD00007200
Molecular Formula: C7H6FNO2
Molecular Weight: 155.13
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 139
Flash Point (°C): 98
Specific Gravity (20/20): 1.28

Formula: C₇H₆FNO₂
Boiling Pt: 139 °C
Melting Pt: 27 °C
Density: 1.28 (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00007200
CAS Number: 446-10-6
EINECS: 207-161-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f01b8e6ff62e92987efd1c9d47b0fb76c31cd20b&vid=d11e715706cc322885912e502ebc8a6c2f8990ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR