(3-Fluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
F0216-5G
F0216-25G
TCF0216-5GEA
73.87
CAD
TCF0216-5G
TCF0216-025G
(3-Fluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)
(3-Fluorophenyl)acetic acid
CAS Number: 331-25-9
MDL Number: MFCD00004331
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 125
Melting point (°C): 47
Flash Point (°C): 110
Formula:
(C₆H₄F)CH₂COOH Boiling Pt: 125 °C Melting Pt: 47 °C Flash Pt: 110 °C |
MDL Number:
MFCD00004331 CAS Number: 331-25-9 EINECS: 206-360-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...