(4-Fluorophenyl)acetic acid ≥98.0%
Supplier: TCI America
Synonyms:
2-(4-Fluorophenyl)acetic acid
F0206-5G
F0206-25G
TCF0206-250GEA
742.09
CAD
TCF0206-250G
TCF0206-005G
TCF0206-025G
(4-Fluorophenyl)acetic acid ≥98.0%
(4-Fluorophenyl)acetic acid
CAS Number: 405-50-5
MDL Number: MFCD00004343
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 164
Melting point (°C): 85
Formula:
(C₆H₄F)CH₂COOH Boiling Pt: 164 °C Melting Pt: 85 °C |
MDL Number:
MFCD00004343 CAS Number: 405-50-5 EINECS: 206-972-4 Merck Index: 13,04204 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...