(2-Fluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(2-Fluorophenyl)acetic acid, 2-Fluorophenylacetic acid

F0205-25G F0205-5G
TCF0205-025GEA 112.18 CAD
TCF0205-025G TCF0205-005G
(2-Fluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)
(2-Fluorophenyl)acetic acid

CAS Number: 451-82-1
MDL Number: MFCD00004315
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 63

Formula: (C₆H₄F)CH₂COOH
MW: 154.14 g/mol
Boiling Pt: 263
Melting Pt: 63 °C
MDL Number: MFCD00004315
CAS Number: 451-82-1
EINECS: 207-196-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e2f0d4f744b0fb7486d65fdcfbc5d141d0a72fc&vid=9e2f0d4f744b0fb7486d65fdcfbc5d141d0a72fc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR