9-Fluorenylmethanol ≥98.0%

Supplier: TCI America

F0200-25G F0200-5G
TCF0200-25GEA 83.74 CAD
TCF0200-25G TCF0200-005G
9-Fluorenylmethanol ≥98.0%
9-Fluorenylmethanol

CAS Number: 24324-17-2
MDL Number: MFCD00001139
Molecular Formula: C14H12O
Molecular Weight: 196.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 105


Other Product Names / Synonyms: 9-Hydroxymethylfluorene

Formula: C₁₄H₁₂O
Boiling Pt: 337 °C (1013 hPa)
Melting Pt: 105 °C
MDL Number: MFCD00001139
CAS Number: 24324-17-2
EINECS: 246-167-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ffa008c9dfaa7d5f93c61e9f05c1644b6bec093&vid=6ffa008c9dfaa7d5f93c61e9f05c1644b6bec093&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR