4-Fluorobenzophenone ≥99.0%

Supplier: TCI America
Warning

F0161-5G F0161-25G
TCF0161-005GEA 26.86 CAD
TCF0161-005G TCF0161-025G
4-Fluorobenzophenone ≥99.0%
4-Fluorobenzophenone

CAS Number: 345-83-5
MDL Number: MFCD00000352
Molecular Formula: C13H9FO
Molecular Weight: 200.21
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 161
Melting point (°C): 49
Flash Point (°C): 113

Formula: C₁₃H₉FO
Boiling Pt: 161 °C
Melting Pt: 49 °C
Flash Pt: 113 °C
MDL Number: MFCD00000352
CAS Number: 345-83-5
EINECS: 206-463-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47eea45688704c6e3acbc9ca6080f8a8f2f9fb6a&vid=47eea45688704c6e3acbc9ca6080f8a8f2f9fb6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR