4-Fluorophenol ≥99.0%

Supplier: TCI America
Warning

Synonyms: p-Fluorophenol, 1-Fluoro-4-hydroxybenzene

F0160-500G F0160-25G
TCF0160-500GEA 483.48 CAD
TCF0160-500G TCF0160-25G
4-Fluorophenol ≥99.0%
4-Fluorophenol

CAS Number: 371-41-5
MDL Number: MFCD00002316
Molecular Formula: C6H5FO
Molecular Weight: 112.10
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 185
Melting point (°C): 47
Flash Point (°C): 88
Storage Temperature: 0-10°C

Formula: C₆H₅FO
MW: 112.1 g/mol
Boiling Pt: 185 °C
Melting Pt: 47 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00002316
CAS Number: 371-41-5
EINECS: 206-736-0
UN: 2923
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1960455824ec284d185642fd05aad078e967b2dc&vid=e2068d4550fbf4f890f2cc6ad8869b6088840f12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR