Formamidine acetate ≥98.0%

Supplier: TCI America

F0154-250G F0154-25G
TCF0154-250GEA 180.12 CAD
TCF0154-250G TCF0154-025G
Formamidine acetate ≥98.0%
Formamidine acetate

CAS Number: 3473-63-0
MDL Number: MFCD00012866
Molecular Formula: CH4N2
Molecular Weight: 104.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 161

Formula: C₃H₈N₂O₂
Melting Pt: 161 °C
MDL Number: MFCD00012866
CAS Number: 3473-63-0
EINECS: 222-442-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=464974b91ea758db28e562a2ab49f360a6953160&vid=464974b91ea758db28e562a2ab49f360a6953160&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR