Fluorescein disodium salt
Supplier: TCI America
Synonyms:
Yellow No. 8
, 2-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium salt
, Disodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate
, 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, disodium salt
, Acid Yellow 73
, Fluorescein sodium salt
CAS Number: 518-47-8
MDL Number: MFCD00167039
Molecular Formula: C20H12O5
Molecular Weight: 376.27
Form: Crystal
Color: Deep Yellow
Other Product Names / Synonyms: Fluorescein Sodium Salt
Formula:
C₂₀H₁₀Na₂O₅ MW: 376.28 g/mol Melting Pt: > 300 °C Density: 1.601 g/cm³ (20 °C) |
MDL Number:
MFCD00167039 CAS Number: 518-47-8 Merck Index: 13,04185 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...