2-Furoic acid 98,0

Supplier: TCI America

Synonyms: Furan-2-carboxylic acid, 2-Furancarboxylic acid

F0081-25G F0081-500G
TCF0081-025GEA 37.11 CAD
TCF0081-025G TCF0081-500G
2-Furoic acid 98,0
2-Furoic acid

CAS Number: 88-14-2
MDL Number: MFCD00003238
Molecular Formula: C5H4O3
Molecular Weight: 112.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 232
Melting point (°C): 132
Flash Point (°C): 137


Other Product Names / Synonyms: 2-Furoic Acid, Pyromucic Acid

Formula: C₅H₄O₃
MW: 112.08 g/mol
Boiling Pt: 232 °C
Melting Pt: 132 °C
Flash Pt: 137 °C
MDL Number: MFCD00003238
CAS Number: 88-14-2
EINECS: 201-803-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf1d79bb506cb52aabd95c87f7e8b3413f444062&vid=bf1d79bb506cb52aabd95c87f7e8b3413f444062&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR