D-(-)-Fructose ≥99.0% (by HPLC)

Supplier: TCI America

F0060-25G F0060-500G
TCF0060-025GEA 44.71 CAD
TCF0060-025G TCF0060-500G
D-(-)-Fructose ≥99.0% (by HPLC)
D(-)-Fructose

CAS Number: 57-48-7
MDL Number: MFCD00148910
Molecular Formula: C6H12O6
Molecular Weight: 180.16
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -92 deg (C=4, H2O)


Product Categories: Bioscience, Basic Sugars (Mono & Oligosaccharides), Fructoses
Other Product Names / Synonyms: D-(-)-Levulose

Formula: C₆H₁₂O₆
Boiling Pt: 552 °C (1013 hPa)
Melting Pt: 100…110 °C
Density: 1.59 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148910
CAS Number: 57-48-7
EINECS: 200-333-3
Merck Index: 13,04295
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3708d5f7545f2c383536c255c43519688b7edc2c&vid=3708d5f7545f2c383536c255c43519688b7edc2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR