FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 3',6'-Dihydroxy-5-isothiocyanato-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 5-FITC, Fluorescein Isothiocyanate, 5-isothiocyanatofluorescein, Fluorescein Isothiocyanate, Isomer I

F0026-100MG F0026-1G
TCF0026-100MGEA 100.8 CAD
TCF0026-100MG TCF0026-1G
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥97.0% (by titrimetric analysis)
FITC (Fluorescein 5-isothiocyanate, Fluorescein isothiocyanate isomer I)

CAS Number: 3326-32-7
Molecular Formula: C21H11NO5S
Molecular Weight: 389.38
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Lambda max.: 499 nmStorage Temperature: 0-10°C


Product Categories: Bioscience, Stains & Dyes, Fluorometry & Fluorescent Imaging, Fluorescent Labeling Dyes, Analytical Science, Fluorescent Dyes, Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: Fluorescein Isothiocyanate, 5-Isothiocyanatofluorescein, 5-FITC

Formula: C₂₁H₁₁NO₅S
MW: 389.381 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00005063
CAS Number: 3326-32-7
EINECS: 222-042-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e457b0d0093d499116cb374ffe3236eccf98a1e9&vid=e457b0d0093d499116cb374ffe3236eccf98a1e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR