Fluorene-9-carboxylic acid ≥97.0%

Supplier: TCI America

Synonyms: 9H-Fluorene-9-carboxylic acid

F0022-5G F0022-25G
TCF0022-005GEA 80.39 CAD
TCF0022-005G TCF0022-025G
Fluorene-9-carboxylic acid ≥97.0%
Fluorene-9-carboxylic acid

CAS Number: 1989-33-9
MDL Number: MFCD00001136
Molecular Formula: C14H10O2
Molecular Weight: 210.23
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 230


Other Product Names / Synonyms: Diphenyleneacetic Acid

Formula: C₁₄H₁₀O₂
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 230 °C
Density: 1.333 g/cm³ (20 °C)
MDL Number: MFCD00001136
CAS Number: 1989-33-9
EINECS: 217-866-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8a7058c3fd9a9cd56d9ef9eada75aa73e8acc4c&vid=e8a7058c3fd9a9cd56d9ef9eada75aa73e8acc4c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR