2-Ethoxybenzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: (2-Ethoxyphenyl)methanol

E0499-25G E0499-5G
TCE0499-025GEA 810.54 CAD
TCE0499-025G TCE0499-5G
2-Ethoxybenzyl alcohol ≥98.0%
2-Ethoxybenzyl alcohol

CAS Number: 71672-75-8
MDL Number: MFCD00004616
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 265
Flash Point (°C): 110
Specific Gravity (20/20): 1.08

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 265
Melting Pt: 0
Density: 1.08 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004616
CAS Number: 71672-75-8
EINECS: 275-811-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00d78456e09e24f935314c971ef8a9eedfb7c770&vid=00d78456e09e24f935314c971ef8a9eedfb7c770&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR