1-Ethyl-3-methylimidazoliumtrifluoromethanesulfonate ≥98.0%

Supplier: TCI America

Synonyms: 1-Ethyl-3-methylimidazolium trifluoromethanesulfonate, 1-Ethyl-3-methylimidazolium trifluoromethanesulphonate

E0494-25G E0494-5G
TCE0494-25GEA 466.13 CAD
TCE0494-25G TCE0494-005G
1-Ethyl-3-methylimidazoliumtrifluoromethanesulfonate ≥98.0%
1-Ethyl-3-methylimidazolium trifluoromethanesulphonate

CAS Number: 145022-44-2
MDL Number: MFCD00210008
Molecular Formula: C7H11F3N2O3S
Molecular Weight: 260.23
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Specific Gravity (20/20): 1.39


Product Categories: Synthetic Organic Chemistry, Ionic Liquids
Other Product Names / Synonyms: 1-Ethyl-3-methylimidazolium Triflate

Formula: C₇H₁₁F₃N₂O₃S
MW: 260.24 g/mol
Density: 1.39 (20 °C)
MDL Number: MFCD00210008
CAS Number: 145022-44-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc49338adbdb7a363ff593c70268bb7db53d726&vid=1bc49338adbdb7a363ff593c70268bb7db53d726&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR