1-Ethyl-3-methylimidazolium chloride ≥98.0%

Supplier: TCI America
Warning

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium chloride

E0490-250G E0490-5G E0490-25G
TCE0490-250GEA 571.96 CAD
TCE0490-250G TCE0490-5G TCE0490-025G
1-Ethyl-3-methylimidazolium chloride ≥98.0%
1-Ethyl-3-methylimidazolium chloride

[for Molten Salt]
CAS Number: 65039-09-0
MDL Number: MFCD00074843
Molecular Formula: C6H11ClN2
Molecular Weight: 146.62
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 84
Flash Point (°C): 186

Formula: C₆H₁₁ClN₂
MW: 146.62 g/mol
Melting Pt: 80
Density: ∼1.1 g/cm³ (20 °C)
Flash Pt: 186
MDL Number: MFCD00074843
CAS Number: 65039-09-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c46c9dfe7cfdd94c57aea649b735b188ea72a2a3&vid=cc0755386a6dec74115de8b54a3910bb02035675&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR