1-Ethyl-2,3-piperazinedione ≥98.0%
Supplier: TCI America
E0459-250G
E0459-25G
TCE0459-250GEA
1652.68
CAD
TCE0459-250G
TCE0459-025G
1-Ethyl-2,3-piperazinedione ≥98.0%
1-Ethyl-2,3-piperazinedione
CAS Number: 59702-31-7
MDL Number: MFCD00051825
Molecular Formula: C6H10N2O2
Molecular Weight: 142.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 125
Other Product Names / Synonyms: 1-Ethyl-2,3-diketopiperazine, 1-Ethyl-2,3-piperazinedione
Formula:
C₆H₁₀N₂O₂ Melting Pt: 125 °C |
MDL Number:
MFCD00051825 CAS Number: 59702-31-7 EINECS: 261-866-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...