(Ethoxycarbonylmethyl)triphenylphosphonium bromide ≥97.0%

Supplier: TCI America

Synonyms: (Carbethoxymethyl)triphenylphosphonium bromide, (2-Ethoxy-2-oxoethyl)triphenylphosphonium bromide

E0407-250G E0407-25G
TCE0407-250GEA 529.3 CAD
TCE0407-250G TCE0407-025G
(Ethoxycarbonylmethyl)triphenylphosphonium bromide ≥97.0%
(Ethoxycarbonylmethyl)triphenylphosphonium bromide

CAS Number: 1530-45-6
MDL Number: MFCD00011835
Molecular Formula: C22H22BrO2P
Molecular Weight: 429.29
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 158


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Carbethoxymethyl(triphenyl)phosphonium Bromide

Formula: C₂₂H₂₂BrO₂P
MW: 429.30 g/mol
Melting Pt: 158 °C
MDL Number: MFCD00011835
CAS Number: 1530-45-6
EINECS: 216-230-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=532a93c77b6a5e51679afd7760b5570c6f4ca520&vid=532a93c77b6a5e51679afd7760b5570c6f4ca520&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR