4-Ethyl-2-methoxyphenol ≥98.0%

Supplier: TCI America

Synonyms: 5-Ethyl-2-hydroxyanisole, 1-Hydroxy-2-methoxy-4-ethyl Benzene, 4-Ethylguaiacol

E0353-5ML E0353-25ML
TCE0353-5MLEA 36.34 CAD
TCE0353-5ML TCE0353-025ML
4-Ethyl-2-methoxyphenol ≥98.0%
4-Ethyl-2-methoxyphenol

CAS Number: 2785-89-9
MDL Number: MFCD00038714
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 235
Flash Point (°C): 108
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: 4-Ethylguaiacol, 4-Ethyl-2-methoxyphenol

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 235 °C
Density: 1.06 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00038714
CAS Number: 2785-89-9
EINECS: 220-500-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0dc4564a3754fa4d791a2e07165d46a9fb35e178&vid=0dc4564a3754fa4d791a2e07165d46a9fb35e178&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR