4'-Ethylacetophenone ≥97.0%

Supplier: TCI America

E0318-25ML E0318-500ML
TCE0318-025MLEA 64.78 CAD
TCE0318-025ML TCE0318-500ML
4'-Ethylacetophenone ≥97.0%
4'-Ethylacetophenone

CAS Number: 937-30-4
MDL Number: MFCD00009262
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 239
Flash Point (°C): 90
Specific Gravity (20/20): 0.99

Formula: C₁₀H₁₂O
Boiling Pt: 239 °C
Density: 0.99 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00009262
CAS Number: 937-30-4
EINECS: 213-326-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c981b5a59c6fe25059d1f09b1cb819e6c09a82ad&vid=c981b5a59c6fe25059d1f09b1cb819e6c09a82ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR