1,2-Epoxydecane ≥97.0%
Supplier: TCI America
E0315-500ML
E0315-25ML
TCE0315-500MLEA
831.08
CAD
TCE0315-500ML
TCE0315-025ML
1,2-Epoxydecane ≥97.0%
1,2-Epoxydecane
CAS Number: 2404-44-6
MDL Number: MFCD00005158
Molecular Formula: C10H20O
Molecular Weight: 156.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Flash Point (°C): 78
Specific Gravity (20/20): 0.84
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2-Decylene Oxide
Formula:
C₁₀H₂₀O Boiling Pt: 208 °C Density: 0.84 (20 °C) Flash Pt: 78 °C |
MDL Number:
MFCD00005158 CAS Number: 2404-44-6 EINECS: 219-295-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...