1,2-Epoxytetradecane ≥95.0%
Supplier: TCI America
|
Warning
|
E0314-25ML
E0314-500ML
TCE0314-025MLEA
64.78
CAD
TCE0314-025ML
TCE0314-500ML
1,2-Epoxytetradecane ≥95.0%
1,2-Epoxytetradecane
CAS Number: 3234-28-4
MDL Number: MFCD00037524
Molecular Formula: C14H28O
Molecular Weight: 212.38
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 113
Specific Gravity (20/20): 0.84
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2-Tetradecylene Oxide
Formula:
C₁₄H₂₈O Boiling Pt: 96 °C Density: 0.84 (20 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037524 CAS Number: 3234-28-4 EINECS: 221-781-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...