1,2-Epoxyoctadecane ≥85.0%

Supplier: TCI America

E0313-25G E0313-400G
TCE0313-025GEA 77.42 CAD
TCE0313-025G TCE0313-400G
1,2-Epoxyoctadecane ≥85.0%
1,2-Epoxyoctadecane

CAS Number: 7390-81-0
MDL Number: MFCD00005152
Molecular Formula: C18H36O
Molecular Weight: 268.49
Purity/Analysis Method: >85.0% (GC)
Form: Crystal
Boiling point (°C): 137
Flash Point (°C): 176
Freezing point (°C): 27


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2-Octadecylene Oxide

Formula: C₁₈H₃₆O
Boiling Pt: 137 °C
Flash Pt: 176 °C
MDL Number: MFCD00005152
CAS Number: 7390-81-0
EINECS: 230-977-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5b90591b45d058251cf05bf69c8c88b94f4850f&vid=e5b90591b45d058251cf05bf69c8c88b94f4850f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR