N-Ethylpyridine-4-methylamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N-(4-Pyridylmethyl)ethylamine, 4-(Ethylaminomethyl)pyridine

E0283-25G E0283-250G
TCE0283-025GEA 110.76 CAD
TCE0283-025G TCE0283-250G
N-Ethylpyridine-4-methylamine ≥97.0% (by GC, titration analysis)
N-Ethylpyridine-4-methylamine

CAS Number: 33403-97-3
MDL Number: MFCD00023632
Molecular Formula: C8H12N2
Molecular Weight: 136.20
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): 98
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: N-Ethyl-4-picolylamine, N-(4-Pyridylmethyl)ethylamine

Formula: C₈H₁₂N₂
MW: 136.20 g/mol
Boiling Pt: 105 °C
Density: 0.99 (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00023632
CAS Number: 33403-97-3
EINECS: 251-506-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c19999bc236b3e77a102c5a51af7b04cf02bff2&vid=8c19999bc236b3e77a102c5a51af7b04cf02bff2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR