Diethyl ethoxymethylenemalonate ≥98.0%

Supplier: TCI America

E0255-500ML E0255-25ML E0255-100ML
TCE0255-500MLEA 164.32 CAD
TCE0255-500ML TCE0255-025ML TCE0255-100ML
Diethyl ethoxymethylenemalonate ≥98.0%
Diethyl ethoxymethylenemalonate

CAS Number: 87-13-8
MDL Number: MFCD00009148
Molecular Formula: C10H16O5
Molecular Weight: 216.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 166
Melting point (°C): -33
Flash Point (°C): 155
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: Ethoxymethylenemalonic Acid Diethyl Ester

Formula: C₁₀H₁₆O₅
Boiling Pt: 166 °C
Melting Pt: –33 °C
Density: 1.08 (20 °C)
Flash Pt: 155 °C
MDL Number: MFCD00009148
CAS Number: 87-13-8
EINECS: 201-725-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=059d9b4e53d344ced383f142e14b1b051589b478&vid=059d9b4e53d344ced383f142e14b1b051589b478&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR