2-Ethyl-4-methylimidazole ≥95.0%
Supplier: TCI America
E0232-25G
E0232-500G
E0232-100G
TCE0232-025GEA
53.72
CAD
TCE0232-025G
TCE0232-500G
TCE0232-100G
2-Ethyl-4-methylimidazole ≥95.0%
2-Ethyl-4-methylimidazole
CAS Number: 931-36-2
MDL Number: MFCD00005193
Molecular Formula: C6H10N2
Molecular Weight: 110.16
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 164
Melting point (°C): 40
Flash Point (°C): 154
Formula:
C₆H₁₀N₂ Boiling Pt: 164 °C Melting Pt: 40 °C Density: 0.97 g/cm³ (20 °C) Flash Pt: 154 °C |
MDL Number:
MFCD00005193 CAS Number: 931-36-2 EINECS: 213-234-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...