2-Ethoxyphenol ≥99.0%
Supplier: TCI America
|
Warning
|
E0230-25G
E0230-500G
TCE0230-025GEA
36.34
CAD
TCE0230-025G
TCE0230-500G
2-Ethoxyphenol ≥99.0%
2-Ethoxyphenol
CAS Number: 94-71-3
MDL Number: MFCD00002187
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 216
Flash Point (°C): 91
Freezing point (°C): 27
Other Product Names / Synonyms: Catechol Monoethyl Ether, Guethol, Pyrocatechol Monoethyl Ether
Formula:
C₈H₁₀O₂ Boiling Pt: 216 °C Melting Pt: 28…29 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002187 CAS Number: 94-71-3 EINECS: 202-358-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...