4-Ethoxyphenol ≥98.0%

Supplier: TCI America
Warning

E0216-25G E0216-500G
TCE0216-025GEA 80.58 CAD
TCE0216-025G TCE0216-500G
4-Ethoxyphenol ≥98.0%
4-Ethoxyphenol

CAS Number: 622-62-8
MDL Number: MFCD00002334
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 121
Melting point (°C): 65


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: Hydroquinone Monoethyl Ether

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 121 °C
Melting Pt: 65 °C
MDL Number: MFCD00002334
CAS Number: 622-62-8
EINECS: 210-748-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR