Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) ≥95.0% (by titrimetric analysis)
Supplier: TCI America
E0194-25G
E0194-500G
E0194-100G
TCE0194-025GEA
82.87
CAD
TCE0194-025G
TCE0194-500G
TCE0194-100G
Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) ≥95.0% (by titrimetric analysis)
Potassium xanthogenate (Potassium O-ethyl dithiocarbonate)
CAS Number: 140-89-6
MDL Number: MFCD00004931
Molecular Formula: C3H6OS2
Molecular Weight: 160.29
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 210
Flash Point (°C): 96
Other Product Names / Synonyms: Ethylxanthic Acid Potassium Salt, Potassium O-Ethyl Dithiocarbonate, Potassium Xanthogenate
Formula:
C₃H₅KOS₂ Melting Pt: 210 °C Flash Pt: 96 °C |
MDL Number:
MFCD00004931 CAS Number: 140-89-6 EINECS: 205-439-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...