Ethylurea ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-Ethylurea

E0192-500G E0192-25G
TCE0192-500GEA 722.06 CAD
TCE0192-500G TCE0192-025G
Ethylurea ≥95.0% (by titrimetric analysis)
Ethylurea

CAS Number: 625-52-5
MDL Number: MFCD00007953
Molecular Formula: C3H8N2O
Molecular Weight: 88.11
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 92
Flash Point (°C): 93

Formula: C₃H₈N₂O
MW: 88.11 g/mol
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 92 °C
Density: 1.213 g/cm³ (18 °C)
Flash Pt: 93 °C
MDL Number: MFCD00007953
CAS Number: 625-52-5
EINECS: 210-898-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1911f70a1dfacc9fa3597ac024be70259189fe20&vid=1911f70a1dfacc9fa3597ac024be70259189fe20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR