5-Ethyl-2-methylpyridine ≥95.0%
Supplier: TCI America
|
Danger
|
E0142-500ML
E0142-25ML
TCE0142-500MLEA
88.43
CAD
TCE0142-500ML
TCE0142-25ML
5-Ethyl-2-methylpyridine ≥95.0%
5-Ethyl-2-methylpyridine
CAS Number: 104-90-5
MDL Number: MFCD00006344
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 178
Flash Point (°C): 68
Freezing point (°C): -70
Specific Gravity (20/20): 0.92
Other Product Names / Synonyms: 5-Ethyl-2-picoline
Formula:
C₈H₁₁N Boiling Pt: 178 °C Melting Pt: –71 °C Density: 0.92 (20 °C) Flash Pt: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006344 CAS Number: 104-90-5 EINECS: 203-250-0 UN: 2300 ADR: 6.1,III Merck Index: 13,03876 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...