(±)-2-Ethyl-1-hexanol ≥99.5%

Supplier: TCI America
Warning

Synonyms: 2-Ethylhexan-1-ol

E0122-25ML E0122-500ML
TCE0122-025MLEA 45.66 CAD
TCE0122-025ML TCE0122-500ML
(±)-2-Ethyl-1-hexanol ≥99.5%
(±)-2-Ethyl-1-hexanol

CAS Number: 104-76-7
MDL Number: MFCD00004746
Molecular Formula: C8H18O
Molecular Weight: 130.23
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 187
Flash Point (°C): 78
Specific Gravity (20/20): 0.83


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 2-Ethylhexyl Alcohol, Octanol, Octyl Alcohol

Formula: CH₃(CH₂)₃CH(C₂H₅)CH₂OH
Boiling Pt: 187 °C
Melting Pt: –76 °C
Density: 0.83 (20 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00004746
CAS Number: 104-76-7
EINECS: 203-234-3
REACH: 01-2119487289-20
Merck Index: 13,03842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6c571ec93bf72509caaa59a0d8afae06060d631&vid=e6c571ec93bf72509caaa59a0d8afae06060d631&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR